Assay Details
Target Gene Details
Entrez Gene ID: | 224648 |
Gene Name: | UHRF1 (ICBP90) binding protein 1 |
Gene Aliases: |
1110020K19Rik, AA408674, F830021D11Rik, mKIAA4127 |
Location: |
Chr.17:27856507-27900040 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uhrf1bp1 | NM_001080769.1 | NP_001074238.1 | ||
AK089790.1 | ||||
AK144200.1 | BAE25764.1 | |||
AK220293.1 | BAD90218.1 |
