Assay Details
Target Gene Details
Entrez Gene ID: | 12385 |
Gene Name: | catenin (cadherin associated protein), alpha 1 |
Gene Aliases: |
2010010M04Rik, AA517462, AI988031, Catna1 |
Location: |
Chr.18:35118912-35254775 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctnna1 | NM_009818.1 | NP_033948.1 | ||
| AK004623.1 | BAB23418.1 | |||
| AK008121.1 | ||||
| AK008181.1 | ||||
| AK008288.1 | ||||
| AK036752.1 | ||||
| AK148496.1 | BAE28588.1 | |||
| AK149721.1 | ||||
| AK149738.1 | ||||
| AK159339.1 | BAE35001.1 | |||
| AK168635.1 | BAE40495.1 | |||
| BC048163.1 | AAH48163.1 | |||
| BC068316.1 | AAH68316.1 | |||
| BC105576.1 | ||||
| D90362.1 | BAA14376.1 | |||
| X59990.1 | CAA42607.1 |
More Information
Set Membership: |
|

Genomic Map