Assay Details
Target Gene Details
Entrez Gene ID: | 110695 |
Gene Name: | aldehyde dehydrogenase family 7, member A1 |
Gene Aliases: |
Atq1, D18Wsu181e |
Location: |
Chr.18:56524722-56572939 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh7a1 | NM_001127338.1 | 7 | 717 | NP_001120810.1 |
| NM_138600.4 | 5 | 523 | NP_613066.2 | |
| AK004991.1 | 7 | 651 | BAB23726.1 | |
| AK145873.1 | 5 | 466 | BAE26715.1 | |
| AK160117.1 | 5 | 450 | BAE35641.1 | |
| AK169195.1 | 5 | 466 | BAE40971.1 | |
| BC012407.1 | 5 | 485 | AAH12407.1 |
More Information
Set Membership: |
|

Genomic Map