Assay Details
Target Gene Details
Entrez Gene ID: | 20532 |
Gene Name: | solute carrier family 3, member 1 |
Gene Aliases: |
D2H, NBAT, NTAA, RBAT |
Location: |
Chr.17:85028347-85064241 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc3a1 | NM_009205.2 | 2 | 564 | NP_033231.2 |
| AB009290.1 | ||||
| AK052623.1 | 2 | 535 | BAC35066.1 | |
| AK132275.1 | 2 | 536 | BAE21075.1 | |
| BC013441.1 | 2 | 506 | AAH13441.1 | |
| CA493293.1 | 2 | 609 | ||
| D88533.1 | 2 | 524 | BAA24427.1 |
More Information
Set Membership: |
|

Genomic Map