Assay Details
Target Gene Details
Entrez Gene ID: | 12558 |
Gene Name: | cadherin 2 |
Gene Aliases: |
CDHN, N-cadherin, Ncad |
Location: |
Chr.18:16588877-16809049 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh2 | NM_007664.4 | 3 | 978 | NP_031690.3 |
| AB008811.1 | 3 | 655 | BAA23549.1 | |
| AK032826.1 | 3 | 418 | BAC28044.1 | |
| AK146981.1 | 3 | 1175 | BAE27584.1 | |
| BC022107.1 | 3 | 409 | AAH22107.1 | |
| M31131.1 | 3 | 655 | AAA37353.1 |
More Information
Set Membership: |
|

Genomic Map