Assay Details
Target Gene Details
Entrez Gene ID: | 433102 |
Gene Name: | surfactant associated 2 |
Gene Aliases: |
EG433102 |
Location: |
Chr.17:35649708-35650569 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sfta2 | NM_001163194.1 | 3 | 367 | NP_001156666.1 |
| AK087178.1 | 3 | 368 |
More Information
Set Membership: |
|

Genomic Map