Assay Details
Target Gene Details
Entrez Gene ID: | 68842 |
Gene Name: | tubby like protein 4 |
Gene Aliases: |
1110057P05Rik, 2210038L17Rik, Tusp |
Location: |
Chr.17:6106827-6240637 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tulp4 | NM_001103181.1 | NP_001096651.1 | ||
| NM_054040.3 | NP_473381.1 | |||
| AF219945.1 | AAF87974.1 | |||
| AK147643.1 | BAE28044.1 | |||
| AK161948.1 | ||||
| AK173165.1 | BAD32443.1 | |||
| BC138024.1 | ||||
| BC138025.1 |

Genomic Map