Assay Details
Target Gene Details
Entrez Gene ID: | 224836 |
Gene Name: | ubiquitin specific peptidase 49 |
Gene Aliases: |
C330046L10Rik, Gm545 |
Location: |
Chr.17:47630690-47684067 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp49 | NM_198421.1 | NP_940813.1 | ||
| AK082872.1 | BAC38663.1 | |||
| BC060712.1 | AAH60712.1 |
More Information
Set Membership: |
|

Genomic Map