Assay Details
Target Gene Details
Entrez Gene ID: | 106861 |
Gene Name: | abhydrolase domain containing 3 |
Gene Aliases: |
AA675331, LABH3 |
Location: |
Chr.18:10644411-10706771 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd3 | NM_134130.1 | NP_598891.1 | ||
| AF429302.1 | AAL27565.1 | |||
| AK134676.1 | BAE22236.1 | |||
| AK144238.1 | BAE25790.1 | |||
| BC026770.1 | AAH26770.1 |
More Information
Set Membership: |
|

Genomic Map