Assay Details
Target Gene Details
Entrez Gene ID: | 20975 |
Gene Name: | synaptojanin 2 |
Gene Aliases: |
AI481647, SJ2, mKIAA0348 |
Location: |
Chr.17:5941280-6045553 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synj2 | NM_001113351.1 | 17 | 2579 | NP_001106822.1 |
| NM_001113352.2 | 18 | 2668 | NP_001106823.1 | |
| NM_001113353.2 | 18 | 2668 | NP_001106824.1 | |
| NM_001290698.1 | 12 | 1696 | NP_001277627.1 | |
| NM_011523.2 | 17 | 2579 | NP_035653.2 | |
| AF041857.1 | 18 | 2669 | AAC40141.1 | |
| AF041858.1 | 7 | 931 | AAC40142.1 | |
| AF041859.1 | 11 | 1460 | AAC40143.1 | |
| AF041860.1 | 4 | 511 | AAC40144.1 | |
| AF041861.1 | AAC40145.1 | |||
| AF041862.1 | 17 | 2524 | AAC40146.1 | |
| AK019694.1 | 12 | 1697 | BAB31837.1 | |
| AK029755.1 | 15 | 2086 | ||
| AK038038.1 | 15 | 1991 | ||
| AK129122.1 | 16 | 2096 | BAC97932.1 | |
| AK142919.1 | 17 | 2300 | ||
| AK148053.1 | 18 | 2406 | ||
| AK161467.1 | 12 | 1680 | BAE36412.1 | |
| AK163512.1 | 16 | 2309 | ||
| BC058749.1 | 18 | 2545 | AAH58749.1 | |
| BC060214.1 | 17 | 2425 | AAH60214.1 |
More Information
Set Membership: |
|

Genomic Map