Assay Details
Target Gene Details
Entrez Gene ID: | 20975 |
Gene Name: | synaptojanin 2 |
Gene Aliases: |
AI481647, SJ2, mKIAA0348 |
Location: |
Chr.17:5941280-6045553 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synj2 | NM_001113351.1 | 18 | 2760 | NP_001106822.1 |
| NM_001113352.2 | 19 | 2849 | NP_001106823.1 | |
| NM_001113353.2 | 19 | 2849 | NP_001106824.1 | |
| NM_001290698.1 | 13 | 1877 | NP_001277627.1 | |
| NM_011523.2 | 18 | 2760 | NP_035653.2 | |
| AF041857.1 | 19 | 2850 | AAC40141.1 | |
| AF041858.1 | 8 | 1112 | AAC40142.1 | |
| AF041859.1 | 12 | 1641 | AAC40143.1 | |
| AF041860.1 | 5 | 692 | AAC40144.1 | |
| AF041861.1 | 2 | 137 | AAC40145.1 | |
| AF041862.1 | 18 | 2705 | AAC40146.1 | |
| AK019694.1 | 13 | 1878 | BAB31837.1 | |
| AK029755.1 | 16 | 2267 | ||
| AK038038.1 | 16 | 2172 | ||
| AK129122.1 | 17 | 2277 | BAC97932.1 | |
| AK142919.1 | 18 | 2481 | ||
| AK148053.1 | 19 | 2587 | ||
| AK161467.1 | 13 | 1861 | BAE36412.1 | |
| AK162329.1 | ||||
| AK163512.1 | ||||
| BC058749.1 | 19 | 2726 | AAH58749.1 | |
| BC060214.1 | 18 | 2606 | AAH60214.1 |
More Information
Set Membership: |
|

Genomic Map