Assay Details
Target Gene Details
Entrez Gene ID: | 20815 |
Gene Name: | serine/arginine-rich protein specific kinase 1 |
Gene Aliases: |
AU017960 |
Location: |
Chr.17:28589590-28622636 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srpk1 | NM_016795.3 | NP_058075.2 | ||
| AB012290.1 | BAA25299.1 | |||
| AJ224115.1 | CAA11833.1 | |||
| AK015754.1 | ||||
| AK151152.1 | BAE30158.1 | |||
| AK161346.1 | BAE36339.1 | |||
| AK164733.1 | 1 | 622 | ||
| BC005707.1 | AAH05707.1 | |||
| BC050761.1 | AAH50761.1 |
More Information
Set Membership: |
|

Genomic Map