Assay Details
Target Gene Details
Entrez Gene ID: | 20975 |
Gene Name: | synaptojanin 2 |
Gene Aliases: |
AI481647, SJ2, mKIAA0348 |
Location: |
Chr.17:5941280-6045553 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synj2 | NM_001113351.1 | 7 | 1164 | NP_001106822.1 |
| NM_001113352.2 | 8 | 1253 | NP_001106823.1 | |
| NM_001113353.2 | 8 | 1253 | NP_001106824.1 | |
| NM_001290698.1 | 2 | 281 | NP_001277627.1 | |
| NM_011523.2 | 7 | 1164 | NP_035653.2 | |
| AF041857.1 | 8 | 1254 | AAC40141.1 | |
| AF041859.1 | 1 | 45 | AAC40143.1 | |
| AF041862.1 | 7 | 1109 | AAC40146.1 | |
| AK014584.1 | 4 | 408 | ||
| AK019694.1 | 2 | 282 | BAB31837.1 | |
| AK029755.1 | 5 | 671 | ||
| AK038038.1 | 5 | 577 | ||
| AK129122.1 | 6 | 681 | BAC97932.1 | |
| AK142919.1 | 7 | 885 | ||
| AK148053.1 | 8 | 992 | ||
| AK161467.1 | 2 | 265 | BAE36412.1 | |
| AK163512.1 | 6 | 894 | ||
| AY823997.1 | AAX24091.1 | |||
| BC058749.1 | 8 | 1130 | AAH58749.1 | |
| BC060214.1 | 7 | 1010 | AAH60214.1 |
More Information
Set Membership: |
|

Genomic Map