Assay Details
Target Gene Details
Entrez Gene ID: | 20975 |
Gene Name: | synaptojanin 2 |
Gene Aliases: |
AI481647, SJ2, mKIAA0348 |
Location: |
Chr.17:5941280-6045553 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synj2 | NM_001113351.1 | 9 | 1369 | NP_001106822.1 |
| NM_001113352.2 | 10 | 1458 | NP_001106823.1 | |
| NM_001113353.2 | 10 | 1458 | NP_001106824.1 | |
| NM_001290698.1 | 4 | 486 | NP_001277627.1 | |
| NM_011523.2 | 9 | 1369 | NP_035653.2 | |
| AF041857.1 | 10 | 1459 | AAC40141.1 | |
| AF041859.1 | 3 | 250 | AAC40143.1 | |
| AF041862.1 | 9 | 1314 | AAC40146.1 | |
| AK014584.1 | 6 | 613 | ||
| AK019694.1 | 4 | 487 | BAB31837.1 | |
| AK029755.1 | 7 | 876 | ||
| AK038038.1 | 7 | 782 | ||
| AK129122.1 | 8 | 886 | BAC97932.1 | |
| AK142919.1 | 9 | 1090 | ||
| AK148053.1 | 10 | 1197 | ||
| AK161467.1 | 4 | 470 | BAE36412.1 | |
| AK163512.1 | 8 | 1099 | ||
| AY823997.1 | AAX24091.1 | |||
| BC058749.1 | 10 | 1335 | AAH58749.1 | |
| BC060214.1 | 9 | 1215 | AAH60214.1 |
More Information
Set Membership: |
|

Genomic Map