Assay Details
Target Gene Details
Entrez Gene ID: | 74355 |
Gene Name: | SMC hinge domain containing 1 |
Gene Aliases: |
4931400A14Rik, AW554188, MommeD1, mKIAA0650 |
Location: |
Chr.17:71344489-71475343 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smchd1 | NM_028887.3 | NP_083163.3 | ||
| AK016419.1 | BAB30222.1 | |||
| AK129181.1 | BAC97991.1 | |||
| BC058618.1 | AAH58618.1 | |||
| BC062946.1 | AAH62946.1 |

Genomic Map