Assay Details
Target Gene Details
Entrez Gene ID: | 108077 |
Gene Name: | superkiller viralicidic activity 2-like (S. cerevisiae) |
Gene Aliases: |
4930534J06Rik, AW214248, Ddx13, SKI, Ski2w |
Location: |
Chr.17:34839226-34850204 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Skiv2l | NM_021337.2 | 27 | 3517 | NP_067312.2 |
| AK029763.1 | 27 | 3506 | BAC26604.1 | |
| AK087810.1 | 23 | 4153 | BAC40010.1 | |
| AK139051.1 | 26 | 3591 | ||
| AK159854.1 | 27 | 3518 | BAE35430.1 | |
| AK181557.1 | ||||
| AK183734.1 | ||||
| AK195584.1 | ||||
| BC023478.1 | 15 | 2121 | AAH23478.1 | |
| BC065999.1 | 27 | 3489 | AAH65999.1 | |
| CF546171.1 | 1 | 94 |

Genomic Map