Assay Details
Target Gene Details
Entrez Gene ID: | 240322 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 19 |
Gene Aliases: |
4831407I23Rik, AU015154, D230034E10Rik |
Location: |
Chr.18:58836143-59053682 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts19 | NM_175506.3 | NP_780715.1 | ||
| AK029178.1 | ||||
| AY135183.1 | AAN10155.1 | |||
| BC150736.1 |
More Information
Set Membership: |
|

Genomic Map