Assay Details
Target Gene Details
Entrez Gene ID: | 54402 |
Gene Name: | serine/threonine kinase 19 |
Gene Aliases: |
G11, RP1 |
Location: |
Chr.17:34823993-34836980 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk19 | NM_019442.3 | NP_062315.1 | ||
| AF099934.1 | AAF26174.1 | |||
| AK146739.1 | BAE27400.1 | |||
| AK168865.1 | BAE40684.1 | |||
| BC022681.1 | AAH22681.1 |
More Information
Set Membership: |
|

Genomic Map