Assay Details
Target Gene Details
Entrez Gene ID: | 20519 |
Gene Name: | solute carrier family 22 (organic cation transporter), member 3 |
Gene Aliases: |
EMT, Oct3, Orct3, Slca22a3 |
Location: |
Chr.17:12419972-12507704 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc22a3 | NM_011395.2 | 3 | 1019 | NP_035525.1 |
| AF078750.1 | 3 | 1019 | AAD20978.1 | |
| AF082566.1 | 3 | 902 | AAD53007.1 | |
| AK143988.1 | 3 | 689 | BAE25648.1 | |
| BC137661.1 | 3 | 894 | ||
| BC137662.1 | 3 | 894 |
More Information
Set Membership: |
|

Genomic Map