Assay Details
Target Gene Details
Entrez Gene ID: | 224807 |
Gene Name: | transmembrane protein 63b |
Gene Aliases: |
BC026370 |
Location: |
Chr.17:45660172-45686254 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem63b | NM_198167.3 | 19 | 1921 | NP_937810.2 |
| AK090200.1 | 19 | 1842 | BAC41131.1 | |
| AK159673.1 | 19 | 1926 | BAE35277.1 | |
| BC026370.1 | 2 | 180 | AAH26370.1 | |
| BC057136.1 | 8 | 884 | AAH57136.1 | |
| BC059283.1 | 12 | 1269 | AAH59283.1 |

Genomic Map