Assay Details
Target Gene Details
Entrez Gene ID: | 269033 |
Gene Name: | RIKEN cDNA 4930503L19 gene |
Gene Aliases: |
9930107F24, BB148262, Lars, Las2 |
Location: |
Chr.18:70452210-70472504 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 269033 |
Gene Name: | RIKEN cDNA 4930503L19 gene |
Gene Aliases: |
9930107F24, BB148262, Lars, Las2 |
Location: |
Chr.18:70452210-70472504 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |