Assay Details
Target Gene Details
Entrez Gene ID: | 74011 |
Gene Name: | solute carrier family 25, member 27 |
Gene Aliases: |
3632410G24Rik, 9430092A03Rik, D530043E16Rik, Ucp4 |
Location: |
Chr.17:43638210-43667194 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a27 | NM_028711.3 | NP_082987.1 | ||
| AB106930.1 | BAC66453.1 | |||
| AK014394.1 | BAB29320.1 | |||
| AK021314.1 | BAB32369.1 | |||
| AK043831.1 | BAC31670.1 | |||
| AK054292.1 | ||||
| BC138993.1 | ||||
| BC138994.1 |
More Information
Set Membership: |
|

Genomic Map