Assay Details
Target Gene Details
Entrez Gene ID: | 103775 |
Gene Name: | solute carrier family 25, member 41 |
Gene Aliases: |
4933406J04Rik, AI507137 |
Location: |
Chr.17:57031454-57042086 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a41 | NM_175333.3 | 2 | 159 | NP_780542.1 |
| AK077111.1 | 2 | 160 | BAC36618.1 | |
| BC115588.1 | 3 | 258 | ||
| BC115589.1 | 2 | 152 | ||
| FM165286.1 | 2 | 101 |
More Information
Set Membership: |
|

Genomic Map