Assay Details
Target Gene Details
Entrez Gene ID: | 109115 |
Gene Name: | suppressor of Ty 3 |
Gene Aliases: |
2310066G22Rik, AI315192, SPT3, SPT3L, Supt3h |
Location: |
Chr.17:44777171-45119294 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Supt3 | NM_178652.2 | 6 | 689 | NP_848767.1 |
| AK050321.1 | 6 | 536 | BAC34187.1 | |
| AK075916.1 | 6 | 689 | BAC36052.1 | |
| AK087935.1 | 6 | 534 | BAC40048.1 |
More Information
Set Membership: |
|

Genomic Map