Assay Details
Target Gene Details
Entrez Gene ID: | 21355 |
Gene Name: | transporter 2, ATP-binding cassette, sub-family B (MDR/TAP) |
Gene Aliases: |
ABC18, AI462429, APT2, Abcb3, Ham-2, Ham2, MTP2, PSF2, RING11, Tap-2, Y1, jas |
Location: |
Chr.17:34204085-34216321 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tap2 | NM_011530.3 | NP_035660.3 | ||
| AK020022.1 | ||||
| AK079667.1 | ||||
| AK145234.1 | ||||
| AK151376.1 | BAE30348.1 | |||
| AK151483.1 | BAE30436.1 | |||
| AK151657.1 | ||||
| AK153287.1 | BAE31870.1 | |||
| AK159200.1 | BAE34894.1 | |||
| AK161091.1 | ||||
| AK169645.1 | ||||
| BC005578.1 | AAH05578.1 | |||
| BC051257.1 | AAH51257.2 | |||
| CT010387.1 | CAJ18594.1 | |||
| KJ650229.1 | ||||
| M90459.1 | AAA39609.1 | |||
| U60087.1 | AAB41970.1 | |||
| U60088.1 | AAB41971.1 | |||
| U60089.1 | AAB41972.1 | |||
| U60090.1 | AAB41973.1 | |||
| U60091.1 | AAB41968.1 | |||
| U60092.1 | AAB41969.1 |
More Information
Set Membership: |
|

Genomic Map