Assay Details
Target Gene Details
Entrez Gene ID: | 20975 |
Gene Name: | synaptojanin 2 |
Gene Aliases: |
AI481647, SJ2, mKIAA0348 |
Location: |
Chr.17:5941280-6045553 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synj2 | NM_001113351.1 | 13 | 1944 | NP_001106822.1 |
| NM_001113352.2 | 14 | 2033 | NP_001106823.1 | |
| NM_001113353.2 | 14 | 2033 | NP_001106824.1 | |
| NM_001290698.1 | 8 | 1061 | NP_001277627.1 | |
| NM_011523.2 | 13 | 1944 | NP_035653.2 | |
| AF041857.1 | 14 | 2034 | AAC40141.1 | |
| AF041858.1 | 3 | 296 | AAC40142.1 | |
| AF041859.1 | 7 | 825 | AAC40143.1 | |
| AF041862.1 | 13 | 1889 | AAC40146.1 | |
| AK014584.1 | 10 | 1188 | ||
| AK019694.1 | 8 | 1062 | BAB31837.1 | |
| AK029755.1 | 11 | 1451 | ||
| AK038038.1 | 11 | 1357 | ||
| AK129122.1 | 12 | 1461 | BAC97932.1 | |
| AK142919.1 | 13 | 1665 | ||
| AK148053.1 | 14 | 1771 | ||
| AK161467.1 | 8 | 1045 | BAE36412.1 | |
| AK163512.1 | 12 | 1674 | ||
| BC058749.1 | 14 | 1910 | AAH58749.1 | |
| BC060214.1 | 13 | 1790 | AAH60214.1 |
More Information
Set Membership: |
|

Genomic Map