Assay Details
Target Gene Details
Entrez Gene ID: | 106952 |
Gene Name: | ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3 |
Gene Aliases: |
Centd3, Drag1, E030006K04Rik, cnt-d3 |
Location: |
Chr.18:37972623-37999533 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arap3 | NM_001205336.1 | 22 | 3442 | NP_001192265.1 |
| NM_139206.2 | 22 | 3242 | NP_631945.2 | |
| AF469621.1 | 21 | 3192 | AAL78677.1 | |
| AF469622.1 | 22 | 3383 | AAL78678.1 | |
| AK220486.1 | 21 | 3671 | BAD90507.1 | |
| BC068145.1 | 22 | 3230 | AAH68145.1 |
More Information
Set Membership: |
|

Genomic Map