Assay Details
Target Gene Details
Entrez Gene ID: | 30948 |
Gene Name: | bridging integrator 1 |
Gene Aliases: |
ALP-1, Amphl, BRAMP-2, SH3P9 |
Location: |
Chr.18:32377217-32435740 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bin1 | NM_001083334.1 | NP_001076803.1 | ||
| NM_009668.2 | NP_033798.1 | |||
| AK184020.1 | ||||
| AK191982.1 | ||||
| BC065160.1 | AAH65160.1 | |||
| CF732768.1 | ||||
| CO426739.1 | ||||
| U60884.1 | AAC52661.1 |

Genomic Map