Assay Details
Target Gene Details
Entrez Gene ID: | 225164 |
Gene Name: | mindbomb homolog 1 (Drosophila) |
Gene Aliases: |
DIP-1, E430019M12Rik, Mib, mKIAA1323 |
Location: |
Chr.18:10725625-10818578 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mib1 | NM_144860.2 | NP_659109.2 | ||
| AK053035.1 | BAC35245.1 | |||
| AK080847.1 | BAC38042.1 | |||
| AK161848.1 | BAE36604.1 | |||
| AY147848.1 | AAN18022.1 | |||
| AY149907.1 | AAN75492.1 | |||
| AY245539.1 | AAO91933.1 | |||
| AY974091.1 | AAX84653.1 | |||
| BC011287.1 | AAH11287.1 | |||
| BC083072.1 | AAH83072.1 | |||
| BC131662.1 |
More Information
Set Membership: |
|

Genomic Map