Assay Details
Target Gene Details
Entrez Gene ID: | 65113 |
Gene Name: | Nedd4 family interacting protein 1 |
Gene Aliases: |
0610010M22Rik, N4wbp5 |
Location: |
Chr.18:38418912-38465190 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndfip1 | NM_022996.1 | NP_075372.1 | ||
| AF220209.1 | AAG44248.1 | |||
| AK050560.1 | BAC34324.1 | |||
| BC026372.1 | AAH26372.1 |

Genomic Map