Assay Details
Target Gene Details
Entrez Gene ID: | 13506 |
Gene Name: | desmocollin 2 |
Gene Aliases: |
AW228162, Dsc2a, Dsc2b |
Location: |
Chr.18:20030633-20059552 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dsc2 | NM_013505.3 | NP_038533.1 | ||
| AK028604.1 | ||||
| AK028636.1 | BAC26041.1 | |||
| AK030333.1 | BAC26907.1 | |||
| AK048547.1 | BAC33371.1 | |||
| AK172168.1 | BAE42862.1 | |||
| BC004663.1 | AAH04663.1 | |||
| BC057867.1 | AAH57867.1 | |||
| L33779.1 | AAA79176.1 | |||
| X73885.1 | CAA52089.1 |
More Information
Set Membership: |
|

Genomic Map