Assay Details
Target Gene Details
Entrez Gene ID: | 107035 |
Gene Name: | F-box protein 38 |
Gene Aliases: |
6030410I24Rik, AU044865, AW214031, Moka, SP329 |
Location: |
Chr.18:62504059-62548858 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxo38 | NM_134136.3 | 19 | 3286 | NP_598897.2 |
| AK031096.1 | 19 | 3236 | BAC27250.1 | |
| AK031347.1 | 19 | 3217 | BAC27358.1 | |
| AK089046.1 | 19 | 3255 | BAC40718.1 | |
| AY267463.1 | 19 | 3216 | AAP31028.1 | |
| BC056348.1 | 19 | 3238 | AAH56348.1 |

Genomic Map