Assay Details
Target Gene Details
Entrez Gene ID: | 212307 |
Gene Name: | microtubule-associated protein, RP/EB family, member 2 |
Gene Aliases: |
AI314113, C820009F03Rik, D18Abb1e, EB1, EB2, RP1 |
Location: |
Chr.18:23752333-23893861 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapre2 | NM_001162941.1 | 4 | 395 | NP_001156413.1 |
| NM_001162942.1 | 4 | 338 | NP_001156414.1 | |
| NM_153058.4 | 3 | 446 | NP_694698.3 | |
| AI325879.1 | 4 | 332 | ||
| AK028823.1 | 4 | 396 | BAC26138.1 | |
| AK038562.1 | 3 | 433 | BAC30045.1 | |
| AK040767.1 | 3 | 421 | BAC30700.1 | |
| AK045493.1 | 3 | 329 | BAC32393.1 | |
| AK045904.1 | 3 | 421 | BAE43326.1 | |
| AK046095.1 | 3 | 388 | ||
| AK046221.1 | 3 | 413 | BAC32643.1 | |
| AK148954.1 | 3 | 421 | BAE28700.1 | |
| AK168834.1 | 5 | 280 | BAE40658.1 | |
| AK192344.1 | ||||
| BC025804.1 | 3 | 393 | AAH25804.1 | |
| BC027056.1 | 3 | 412 | AAH27056.1 | |
| BC028987.1 | 3 | 368 | AAH28987.1 | |
| BC035254.1 | 3 | 398 | AAH35254.1 |
More Information
Set Membership: |
|

Genomic Map