Assay Details
Target Gene Details
Entrez Gene ID: | 320253 |
Gene Name: | membrane-associated ring finger (C3HC4) 3 |
Gene Aliases: |
6330411I15Rik, A530081L18Rik, AI843639, MARCH-III |
Location: |
Chr.18:56761716-56925652 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| March3 | NM_177115.2 | NP_796089.1 | ||
| AK041088.1 | BAC30814.1 | |||
| BC107232.1 | AAI07233.1 | |||
| BC107233.1 | AAI07234.1 |
More Information
Set Membership: |
|

Genomic Map