Assay Details
Target Gene Details
Entrez Gene ID: | 19123 |
Gene Name: | protein C |
Gene Aliases: |
PC |
Location: |
Chr.18:32123126-32139570 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Proc | NM_001042767.2 | NP_001036232.1 | ||
| NM_001042768.2 | NP_001036233.2 | |||
| NM_008934.3 | NP_032960.3 | |||
| NR_045173.1 | ||||
| AF318182.1 | AAK07918.1 | |||
| AI118438.1 | ||||
| BC013896.1 | AAH13896.1 | |||
| CA468822.1 | ||||
| CF550816.1 | ||||
| D10445.1 | BAA01235.1 | |||
| DV662175.1 |
More Information
Set Membership: |
|

Genomic Map