Assay Details
Target Gene Details
Entrez Gene ID: | 225341 |
Gene Name: | LIM and senescent cell antigen like domains 2 |
Gene Aliases: |
PINCH2 |
Location: |
Chr.18:31922277-31958620 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lims2 | NM_144862.3 | NP_659111.1 | ||
| BC010816.1 | AAH10816.1 |
More Information
Set Membership: |
|

Genomic Map