Assay Details
Target Gene Details
Entrez Gene ID: | 27528 |
Gene Name: | neuronal regeneration related protein |
Gene Aliases: |
AI325076, D0H4S114, Harp, P311, PTZ17, SEZ17 |
Location: |
Chr.18:33437019-33464029 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrep | NM_001109988.1 | NP_001103458.1 | ||
| AK020629.1 | BAB32157.1 | |||
| BY150793.1 | 1 | 105 | ||
| CJ148521.1 | ||||
| CJ165393.1 |
More Information
Set Membership: |
|

Genomic Map