Assay Details
Target Gene Details
Entrez Gene ID: | 332309 |
Gene Name: | glutaredoxin, cysteine rich 2 |
Gene Aliases: |
Gm851 |
Location: |
Chr.18:41986201-41999049 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grxcr2 | NM_001033426.2 | NP_001028598.1 | ||
| AK158418.1 | BAE34497.1 | |||
| BC147102.1 | ||||
| BC147103.1 |
More Information
Set Membership: |
|

Genomic Map