Assay Details
Target Gene Details
Entrez Gene ID: | 225363 |
Gene Name: | eukaryotic translation termination factor 1 |
Gene Aliases: |
AI463371, D6Ertd109e, ERF, ERF1, SUP45L1, TB3-1 |
Location: |
Chr.18:34902785-34932003 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Etf1 | NM_144866.3 | NP_659115.3 | ||
| AK049607.1 | BAC33839.1 | |||
| AK077559.1 | ||||
| AK139933.1 | ||||
| AK151028.1 | ||||
| AK151710.1 | ||||
| AK152428.1 | BAE31210.1 | |||
| AK152952.1 | BAE31619.1 | |||
| AK164020.1 | BAE37589.1 | |||
| AK166740.1 | BAE38983.1 | |||
| AK169345.1 | BAE41096.1 | |||
| AK170221.1 | BAE41644.1 | |||
| BC013717.1 | AAH13717.1 | |||
| D87691.1 | BAA13439.1 |
More Information
Set Membership: |
|

Genomic Map