Assay Details
Target Gene Details
Entrez Gene ID: | 67222 |
Gene Name: | serum response factor binding protein 1 |
Gene Aliases: |
2810036K01Rik, p49/STRAP |
Location: |
Chr.18:52465672-52490738 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srfbp1 | NM_026040.3 | NP_080316.3 | ||
| AK012865.1 | BAB28520.1 | |||
| AY611629.1 | AAU25828.1 |
More Information
Set Membership: |
|

Genomic Map