Assay Details
Target Gene Details
Entrez Gene ID: | 277250 |
Gene Name: | KDM3B lysine (K)-specific demethylase 3B |
Gene Aliases: |
5830462I21Rik, JHDM2B, Jmjd1b, mKIAA1082 |
Location: |
Chr.18:34776243-34839370 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm3b | NM_001081256.1 | 17 | 4445 | NP_001074725.1 |
| AK033343.1 | 3 | 369 | BAC28239.1 | |
| AK044708.1 | ||||
| AK129281.1 | BAC98091.1 | |||
| AK159090.1 | 4 | 778 | ||
| BC031981.1 | 3 | 514 | AAH31981.1 | |
| BC038376.1 | 7 | 1263 | AAH38376.1 | |
| BC060727.1 | 10 | 2005 | AAH60727.1 | |
| BC108415.1 | 10 | 1849 | AAI08416.1 | |
| BC151084.1 | 17 | 4406 |
More Information
Set Membership: |
|

Genomic Map