Assay Details
Target Gene Details
Entrez Gene ID: | 13511 |
Gene Name: | desmoglein 2 |
Gene Aliases: |
AA408168, D18Ertd293e |
Location: |
Chr.18:20558116-20604526 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dsg2 | NM_007883.2 | NP_031909.2 | ||
| AB072269.1 | BAB86843.1 | |||
| BC020144.1 | AAH20144.1 | |||
| BC044848.1 | AAH44848.1 | |||
| BC131653.1 | ||||
| BC131654.1 |
More Information
Set Membership: |
|

Genomic Map