Assay Details
Target Gene Details
Entrez Gene ID: | 13511 |
Gene Name: | desmoglein 2 |
Gene Aliases: |
AA408168, D18Ertd293e |
Location: |
Chr.18:20558116-20604526 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dsg2 | NM_007883.2 | 8 | 1146 | NP_031909.2 |
| AB072269.1 | 8 | 1174 | BAB86843.1 | |
| BC044848.1 | 8 | 1107 | AAH44848.1 | |
| BC131653.1 | 8 | 1122 | ||
| BC131654.1 | 8 | 1122 |
More Information
Set Membership: |
|

Genomic Map