Assay Details
Target Gene Details
Entrez Gene ID: | 277250 |
Gene Name: | KDM3B lysine (K)-specific demethylase 3B |
Gene Aliases: |
5830462I21Rik, JHDM2B, Jmjd1b, mKIAA1082 |
Location: |
Chr.18:34776243-34839370 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm3b | NM_001081256.1 | NP_001074725.1 | ||
| AK033343.1 | BAC28239.1 | |||
| AK044708.1 | 2 | 556 | ||
| AK129281.1 | BAC98091.1 | |||
| AK159090.1 | ||||
| BC031981.1 | AAH31981.1 | |||
| BC038376.1 | AAH38376.1 | |||
| BC060727.1 | AAH60727.1 | |||
| BC108415.1 | AAI08416.1 | |||
| BC151084.1 |
More Information
Set Membership: |
|

Genomic Map