Assay Details
Target Gene Details
Entrez Gene ID: | 19330 |
Gene Name: | RAB18, member RAS oncogene family |
Gene Aliases: |
AA959686 |
Location: |
Chr.18:6765014-6791606 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab18 | NM_001278447.1 | NP_001265376.1 | ||
| NM_181070.6 | NP_851415.1 | |||
| AB232612.1 | BAF02874.1 | |||
| AK045514.1 | BAC32402.1 | |||
| AK140901.1 | BAE24512.1 | |||
| AK143673.1 | BAE25489.1 | |||
| AK146177.1 | BAE26955.1 | |||
| AK146397.1 | BAE27140.1 | |||
| AK154920.1 | BAE32926.1 | |||
| BC056351.1 | AAH56351.1 | |||
| L04966.1 | AAC37632.1 | |||
| M79308.1 | AAK14832.1 | |||
| X80333.1 | CAA56583.1 |

Genomic Map