Assay Details
Target Gene Details
Entrez Gene ID: | 18169 |
Gene Name: | neuropeptide Y receptor Y6 |
Gene Aliases: |
- |
Location: |
Chr.18:44270127-44277700 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npy6r | NM_010935.3 | 1 | 58 | NP_035065.1 |
| AK030279.1 | 1 | 60 | BAC26875.1 |
More Information
Set Membership: |
|

Genomic Map