Assay Details
Target Gene Details
Entrez Gene ID: | 72512 |
Gene Name: | transmembrane protein 173 |
Gene Aliases: |
2610307O08Rik, ERIS, MPYS, Mita, STING |
Location: |
Chr.18:35733678-35740554 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem173 | NM_001289591.1 | 1 | 85 | NP_001276520.1 |
| NM_001289592.1 | 1 | 85 | NP_001276521.1 | |
| NM_028261.1 | 1 | 85 | NP_082537.1 | |
| AK012006.1 | BAB27972.1 | |||
| AK077788.1 | BAC37010.1 | |||
| AK089405.1 | 1 | 63 | BAC40870.1 | |
| AK157370.1 | BAE34068.1 | |||
| AK158458.1 | BAE34517.1 | |||
| AK170724.1 | 1 | 63 | BAE41981.1 | |
| AK171065.1 | BAE42224.1 | |||
| AK171203.1 | BAE42310.1 | |||
| AK171612.1 | BAE42563.1 | |||
| AX775911.1 | CAE11655.1 | |||
| BB871885.1 | 1 | 87 | ||
| BB872463.1 | 1 | 87 | ||
| BC046640.1 | AAH46640.1 |

Genomic Map