Assay Details
Target Gene Details
Entrez Gene ID: | 66810 |
Gene Name: | RNA binding motif protein 22 |
Gene Aliases: |
8430430L24Rik |
Location: |
Chr.18:60560786-60572729 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm22 | NM_025776.2 | NP_080052.1 | ||
| AK018452.1 | BAB31220.1 | |||
| AK045116.1 | BAC32229.1 | |||
| AK150271.1 | BAE29427.1 | |||
| AK151758.1 | BAE30665.1 | |||
| AK152252.1 | BAE31073.1 | |||
| AK165277.1 | ||||
| AK166600.1 | BAE38885.1 | |||
| AK167509.1 | BAE39585.1 | |||
| AK168919.1 | BAE40732.1 | |||
| BC080205.1 | AAH80205.1 |
More Information
Set Membership: |
|

Genomic Map