Assay Details
Target Gene Details
Entrez Gene ID: | 72512 |
Gene Name: | transmembrane protein 173 |
Gene Aliases: |
2610307O08Rik, ERIS, MPYS, Mita, STING |
Location: |
Chr.18:35733678-35740554 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem173 | NM_001289591.1 | 5 | 921 | NP_001276520.1 |
| NM_001289592.1 | 6 | 888 | NP_001276521.1 | |
| NM_028261.1 | 6 | 1011 | NP_082537.1 | |
| AK012006.1 | 6 | 958 | BAB27972.1 | |
| AK077788.1 | 6 | 912 | BAC37010.1 | |
| AK089405.1 | 6 | 989 | BAC40870.1 | |
| AK146284.1 | 6 | 884 | BAE27042.1 | |
| AK153868.1 | 6 | 882 | BAE32222.1 | |
| AK157370.1 | 5 | 823 | BAE34068.1 | |
| AK158458.1 | 6 | 797 | BAE34517.1 | |
| AK170724.1 | 6 | 989 | BAE41981.1 | |
| AK171065.1 | 6 | 975 | BAE42224.1 | |
| AK171203.1 | 6 | 975 | BAE42310.1 | |
| AK171612.1 | 6 | 942 | BAE42563.1 | |
| AK172314.1 | 5 | 753 | ||
| AX775911.1 | 6 | 898 | CAE11655.1 | |
| BC027757.1 | 4 | 655 | AAH27757.1 | |
| BC046640.1 | 5 | 846 | AAH46640.1 | |
| DQ910493.1 | 6 | 809 | ||
| FJ222242.1 | 4 | 673 |

Genomic Map