Assay Details
Target Gene Details
Entrez Gene ID: | 170947 |
Gene Name: | myozenin 3 |
Gene Aliases: |
4833419K08Rik, Fatz-3 |
Location: |
Chr.18:60573717-60591716 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myoz3 | NM_133363.3 | NP_579941.2 | ||
| AF480442.1 | AAL79335.1 | |||
| BC104046.2 | AAI04047.1 | |||
| BC104047.1 | AAI04048.1 |
More Information
Set Membership: |
|

Genomic Map